跳转至内容
Merck
  • Ecdysone control of developmental transitions: lessons from Drosophila research.
Ecdysone control of developmental transitions: lessons from Drosophila research.
Annual review of entomology (2012-10-18)
Naoki Yamanaka, Kim F Rewitz, Michael B O'Connor
摘要
The steroid hormone ecdysone is the central regulator of insect developmental transitions. Recent new advances in our understanding of ecdysone action have relied heavily on the application of Drosophila melanogaster molecular genetic tools to study insect metamorphosis. In this review, we focus on three major aspects of Drosophila ecdysone biology: (a) factors that regulate the timing of ecdysone release, (b) molecular basis of stage- and tissue-specific responses to ecdysone, and (c) feedback regulation and coordination of ecdysone signaling.
材料
货号
品牌
产品描述
Sigma-Aldrich
α-蜕皮素, ≥90%