跳转至内容
Merck
  • [Effect of pravastatin on transportation of scutellarin in mouse liver and its mechanism].
[Effect of pravastatin on transportation of scutellarin in mouse liver and its mechanism].
Yao xue xue bao = Acta pharmaceutica Sinica (2011-06-02)
Jian-Ming Liu, Yu-Qing Xiong
摘要
This study is to investigate the transportation of scutellarin in cell and live models and study on mechanism of absorption and transport of scutellarin in mouse liver. The concentration of scutellarin in plasma and liver from control and pretreated groups was determined by high performance liquid chromatography. The uptake of scutellarin was examined in control hepatocytes group, induced hepatocytes group and induced hepatocytes plus pravastatin group. Pravastatin can affect the pharmacokinetics of scutellarin in mouse: CL is decreased while AUC is increased. The scutellarin absorption of hepatocyte induced group was higher than that of control group, but was decreased in the group with pravastatin added. The research showed that there was potential drug interaction between pravastatin and scutellarin. The drugs may compete for oatp2 mediated transport pathway consisted in the uptake of scutellarin in liver.
材料
货号
品牌
产品描述
Sigma-Aldrich
5-Pregnen-3β-ol-20-one-16α-carbonitrile, ≥97%