跳转至内容
Merck
  • Ischemia-reperfusion injury of rabbit ovary and protective effect of trapidil: an experimental study.
Ischemia-reperfusion injury of rabbit ovary and protective effect of trapidil: an experimental study.
Pediatric surgery international (2007-12-07)
Salih Somuncu, Murat Cakmak, Gunnur Dikmen, Hülya Akman, Murat Kaya
摘要
We aimed to detect the protective effect of trapidil in ischemia-reperfusion (IR) injury due to ovarian torsion and detorsion. Thirty-two pubertal New Zealand albino rabbits were used. Adnexal torsion was created by rotating the left adnexa including the tubal and ovarian vessels in a 360 degrees clockwise direction. Adnexal detorsion was done by untwisting the adnexa. In the IR group, left oopherectomy was performed after 3 h of adnexal torsion and 3 h of adnexal detorsion. In the study group, a 3-h adnexal torsion was performed and trapidil was administered intraperitoneally as a single dose of 40 mg/kg, 1 h before detorsion. The left oopherectomy was performed after a 3-h adnexal detorsion. In the sham group, sham operation was performed followed by left oopherectomy. In the control group, normal ovarian tissue was evaluated. Catalase, superoxide dismutase (SOD), glutathione peroxidase (GSH-Px), and malondialdehyde (MDA) levels of ovarian tissue were determined for each group. The values of SOD and GSH-Px activities in the IR group were significantly decreased (P < 0.05). In addition, the MDA level was significantly higher in the IR group (P < 0.01). The trapidil-administered group showed significant increase in the levels of GSH-Px (P < 0.05), catalase (P < 0.05), SOD (P < 0.05), and decreased MDA levels (P < 0.05) compared to those in the IR group. The study has shown that trapidil treatment prevents ischemia induced oxidative damage in the ovarian tissues of rabbits.
材料
货号
品牌
产品描述
曲匹地尔, European Pharmacopoeia (EP) Reference Standard