跳转至内容
Merck
  • Altered pharmacokinetics and hepatic uptake of TBuMA in ethynylestradiol-induced cholestasis.
Altered pharmacokinetics and hepatic uptake of TBuMA in ethynylestradiol-induced cholestasis.
Archives of pharmacal research (2006-05-10)
Soon-Sun Hong, Jong-Moon Choi, Hyo-Eon Jin, Chang-Koo Shim
摘要
The objective of this study was to examine the pharmacokinetics of organic cations in intrahepatic cholestatic rats. A pretreatment with 17alpha-ethynylestradiol was used to induce intrahepatic cholestasis, and tributylmethylammonium (TBuMA) was used as a representative model organic cation. When [3H]TBuMA was intravenously administered1 the AUC value for TBuMA was significantly increased by 79% in cholestasis, and its total systemic clearance was consequently decreased by 46%. In addition, the in vivo hepatic uptake clearance of TBuMA from the plasma to the liver was decreased by 50% in cholestasis. The concentration of bile salts in plasma was increased by 2.1 fold in cholestatic rats. Since TBuMA forms ion-pair complexes with anionic components such as bile salts, the decreased hepatic uptake of TBuMA in cholestasis may be due to a change in endogenous components, e.g., bile salts in the plasma. In isolated normal hepatocytes, the uptake clearance for TBuMA in the presence of cholestatic plasma was decreased by 20% compared with normal plasma. Therefore, we conclude that the inhibition of the hepatic uptake process by the cholestasis may be in part due to the increased formation of ion-pair complexes of TBuMA with bile salts in the plasma.
材料
货号
品牌
产品描述
Sigma-Aldrich
三丁基甲基氯化铵, ≥98.0% (T)
Sigma-Aldrich
三丁基甲基氯化铵 溶液, 75 wt. % in H2O