跳转至内容
Merck
  • Nifedipine loaded chitosan microspheres prepared by emulsification phase-separation.
Nifedipine loaded chitosan microspheres prepared by emulsification phase-separation.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (2004-03-03)
S Dhawan, A K Singla
摘要
A high yield of nifedipine-chitosan microspheres could be obtained using an emulsification phase-separation method. A high level of entrapment of nifedipine in the microspheres was achieved. The microspheres exhibited excellent swelling properties. Differential scanning calorimetry, X-ray diffractometry, and scanning electron microscopy confirmed that at 1.84% loading, nifedipine was dispersed molecularly. The microspheres exhibited faster release at low loadings compared to high loadings. Fitting the data to the coupled Fickian/case II equation, showed that at low loadings polymer relaxation coefficients (k2) were high. As the polymer content increased in the microspheres, the value of n (diffusional exponent characteristic of the release mechanism) approached one, which is indicative of zero order.
材料
货号
品牌
产品描述
Sigma-Aldrich
Span ® 85