跳转至内容
Merck
  • Viral strategies for targeting spinal neuronal subtypes in adult wild-type rodents.
Viral strategies for targeting spinal neuronal subtypes in adult wild-type rodents.
Scientific reports (2022-05-24)
Jaspreet Kaur, Rune W Berg
摘要
Targeting specific subtypes of interneurons in the spinal cord is primarily restricted to a small group of genetic model animals. Since the development of new transgenic model animals can be expensive and labor intensive, it is often difficult to generalize these findings and verify them in other model organisms, such as the rat, ferret or monkey, that may be more beneficial in certain experimental investigations. Nevertheless, endogenous enhancers and promoters delivered using an adeno-associated virus (AAV) have been successful in providing expression in specific subtypes of neurons in the forebrain of wildtype animals, and therefore may introduce a shortcut. GABAergic interneurons, for instance, have successfully been targeted using the mDlx promoter, which has recently been developed and is now widely used in wild type animals. Here, we test the specificity and efficiency of the mDlx enhancer for robust targeting of inhibitory interneurons in the lumbar spinal cord of wild-type rats using AAV serotype 2 (AAV2). Since this has rarely been done in the spinal cord, we also test the expression and specificity of the CamKIIa and hSynapsin promoters using serotype 9. We found that AAV2-mDlx does in fact target many neurons that contain an enzyme for catalyzing GABA, the GAD-65, with high specificity and a small fraction of neurons containing an isoform, GAD-67. Expression was also seen in some motor neurons although with low correlation. Viral injections using the CamKIIa enhancer via AAV9 infected in some glutamatergic neurons, but also GABAergic neurons, whereas hSynapsin via AAV9 targets almost all the neurons in the lumbar spinal cord.
材料
货号
品牌
产品描述
Sigma-Aldrich
抗胆碱乙酰基转移酶抗体, Chemicon®, from goat
Sigma-Aldrich
抗-β-肌动蛋白−过氧化物酶抗体,小鼠单克隆 小鼠抗, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
抗谷氨酸受体2抗体,细胞外,克隆6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
抗-GluR1抗体,克隆C3T,兔单克隆, culture supernatant, clone C3T, Upstate®
Sigma-Aldrich
抗谷氨酸脱羧酶65(5-22) 兔抗, IgG fraction of antiserum, buffered aqueous solution