跳转至内容
Merck
  • Multifamily determination of pesticide residues in soya-based nutraceutical products by GC/MS-MS.
Multifamily determination of pesticide residues in soya-based nutraceutical products by GC/MS-MS.
Food chemistry (2014-12-04)
Agneša Páleníková, Gerardo Martínez-Domínguez, Francisco Javier Arrebola, Roberto Romero-González, Svetlana Hrouzková, Antonia Garrido Frenich
摘要
An analytical method based on a modified QuEChERS extraction coupled with gas chromatography-tandem mass spectrometry (GC-MS/MS) was evaluated for the determination of 177 pesticides in soya-based nutraceutical products. The QuEChERS method was optimised and different extraction solvents and clean-up approaches were tested, obtaining the most efficient conditions with a mixture of sorbents (PSA, C18, GBC and Zr-Sep(+)). Recoveries were evaluated at 10, 50 and 100 μg/kg and ranged between 70% and 120%. Precision was expressed as relative standard deviation (RSD), and it was evaluated for more than 160 pesticides as intra and inter-day precision, with values always below 20% and 25%, respectively. Limits of detection (LODs) ranged from 0.1 to 10 μg/kg, whereas limits of quantification (LOQs) from 0.5 to 20 μg/kg. The applicability of the method was proved by analysing soya-based nutraceuticals. Two pesticides were found in these samples, malathion and pyriproxyfen, at 11.1 and 1.5 μg/kg respectively.
材料
货号
品牌
产品描述
Supelco
环草敌, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland