跳转至内容
Merck
  • Protein kinase C-mediated Ca2+ entry in HEK 293 cells transiently expressing human TRPV4.
Protein kinase C-mediated Ca2+ entry in HEK 293 cells transiently expressing human TRPV4.
British journal of pharmacology (2003-09-13)
Feng Xu, Eisaku Satoh, Toshihiko Iijima
摘要
1. We investigated whether protein kinase C (PKC) activation stimulates Ca2+ entry in HEK 293 cells transfected with human TRPV4 cDNA and loaded with fura-2. 2. Phorbol 12-myristate 13-acetate (PMA), a PKC-activating phorbol ester, increased the intracellular Ca2+ concentration ([Ca2+]i) in a dose-dependent manner, with an EC50 value of 11.7 nm. Exposure to a hypotonic solution (HTS) after PMA further increased [Ca2+]i. Two other PKC-activating phorbol esters, phorbol 12,13-didecanoate (PDD) and phorbol 12,13-dibutyrate, also caused [Ca2+]i to increase. 3. The inactive isomer 4alpha-PMA was less effective and the peak [Ca2+]i increase was significantly smaller than that induced by PMA. In contrast, 4alpha-PDD produced a monophasic or biphasic [Ca2+]i increase with a different latency, while 4alpha-phorbol had no effect. 4. The PMA-induced [Ca2+]i increase was abolished by prior exposure to bisindolylmaleimide (BIM), a PKC-specific inhibitor, and suppressed by the nonspecific PKC inhibitor 1-(5-isoquinolinesulphonyl)-2-methylpiperazine. The [Ca2+]i increase induced by 4alpha-PMA, 4alpha-PDD or HTS was not significantly affected by BIM. 5. These results suggest that both PKC-dependent and -independent mechanisms are involved in the phorbol ester-induced activation of TRPV4, and the PKC-independent pathway is predominant in HTS-induced Ca2+ entry.
材料
货号
品牌
产品描述
Sigma-Aldrich
4α-佛波醇12-十四酸酯13-乙酸酯, solid, ≥95% (TLC)