跳转至内容
Merck
  • WS-7338, new endothelin receptor antagonists isolated from Streptomyces sp. No. 7338. I. Taxonomy, fermentation, isolation, physico-chemical properties and biological activities.
WS-7338, new endothelin receptor antagonists isolated from Streptomyces sp. No. 7338. I. Taxonomy, fermentation, isolation, physico-chemical properties and biological activities.
The Journal of antibiotics (1992-01-01)
S Miyata, M Hashimoto, Y Masui, M Ezaki, S Takase, M Nishikawa, S Kiyoto, M Okuhara, M Kohsaka
摘要
WS-7338 A, B, C and D, novel endothelin receptor antagonists, have been isolated from fermentation broth of Streptomyces sp. No. 7338. These antagonists were purified from the culture mycelium by extraction with acetone, followed by carbon column chromatography and HPLC. Among them, WS-7338 B showed good activity in an endothelin receptor binding assay with an IC50 of 2.7 x 10(-7) M.
材料
货号
品牌
产品描述
Sigma-Aldrich
Endothelin Receptor Antagonist W 7338A