跳转至内容
Merck
  • Overexpression of SmANS Enhances Anthocyanin Accumulation and Alters Phenolic Acids Content in Salvia miltiorrhiza and Salvia miltiorrhiza Bge f. alba Plantlets.
Overexpression of SmANS Enhances Anthocyanin Accumulation and Alters Phenolic Acids Content in Salvia miltiorrhiza and Salvia miltiorrhiza Bge f. alba Plantlets.
International journal of molecular sciences (2019-05-09)
Hongyan Li, Jingling Liu, Tianlin Pei, Zhenqing Bai, Ruilian Han, Zongsuo Liang
摘要
Flavonoids play multiple roles in plant coloration and stress resistance and are closely associated with human health. Flavonoids and non-flavonoids (such as phenolic acids) are produced via the phenylpropanoid-derived pathway. Anthocyanidin synthase (ANS) catalyzes the synthesis of anthocyanins from leucoanthocyanidin in the flavonoids branched pathway. In this study, SmANS from Salvia miltiorrhiza was cloned and mainly localized in the endoplasmic reticulum (ER), plastids, Golgi, plasma membrane, and nucleus of tobacco epidermal cells, and was most highly expressed in purple petals in S. miltiorrhiza, whereas it showed almost no expression in white petals, green calyxes, and pistils in S. miltiorrhiza Bge f. alba. Overexpressed SmANS enhanced anthocyanin accumulation but reduced salvianolic acid B (SAB) and rosmarinic acid (RA) biosynthesis in S. miltiorrhiza and S. miltiorrhiza Bge f. alba plantlets, meanwhile, it restored the purple-red phenotype in S. miltiorrhiza Bge f. alba. These changes were due to reallocation of the metabolic flow, which was influenced by the SmANS gene. These findings indicate that SmANS not only plays a key role in anthocyanin accumulation in S. miltiorrhiza, but also acts as a "switch" for the coloration of S. miltiorrhiza Bge f. alba. This study provides baseline information for further research on flavonoids metabolism and improvement of anthocyanin or phenolic acid production by genetic engineering.