跳转至内容
Merck
  • One-Pot Method of Synthesizing TEMPO-Oxidized Bacterial Cellulose Nanofibers Using Immobilized TEMPO for Skincare Applications.
One-Pot Method of Synthesizing TEMPO-Oxidized Bacterial Cellulose Nanofibers Using Immobilized TEMPO for Skincare Applications.
Polymers (2019-06-15)
Seung-Hyun Jun, Sun-Gyoo Park, Nae-Gyu Kang
摘要
In the skincare field, water-dispersed bacterial cellulose nanofibers synthesized via an oxidation reaction using 2,2,6,6-tetramethyl-1-piperidine-N-oxy radical (TEMPO) as a catalyst are promising bio-based polymers for engineered green materials because of their unique properties when applied to the surface of the skin, such as a high tensile strength, high water-holding capacity, and ability to block harmful substances. However, the conventional method of synthesizing TEMPO-oxidized bacterial cellulose nanofibers (TOCNs) is difficult to scale due to limitations in the centrifuge equipment when treating large amounts of reactant. To address this, we propose a one-pot TOCN synthesis method involving TEMPO immobilized on silica beads that employs simple filtration instead of centrifugation after the oxidation reaction. A comparison of the structural and physical properties of the TOCNs obtained via the proposed and conventional methods found similar properties in each. Therefore, it is anticipated that due to its simplicity, efficiency, and ease of use, the proposed one-pot synthesis method will be employed in production scenarios to prepare production quantities of bio-based polymer nanofibers in various potential industrial applications in the fields of skincare and biomedical research.