跳转至内容
Merck
  • m6A methylation modulates adipogenesis through JAK2-STAT3-C/EBPβ signaling.
m6A methylation modulates adipogenesis through JAK2-STAT3-C/EBPβ signaling.
Biochimica et biophysica acta. Gene regulatory mechanisms (2019-07-12)
Ruifan Wu, Guanqun Guo, Zhen Bi, Youhua Liu, Yuanling Zhao, Nana Chen, Fengqin Wang, Yizhen Wang, Xinxia Wang
摘要
N6-methyladenosine (m6A), the most abundant internal mRNA modification in eukaryotes, plays a vital role in regulating adipogenesis. However, its underlying mechanism remains largely unknown. Here, we reveal that deletion of m6A demethylase FTO in porcine and mouse preadipocytes inhibits adipogenesis through JAK2-STAT3-C/EBPβ signaling. Mechanistically, FTO deficiency suppresses JAK2 expression and STAT3 phosphorylation, leading to attenuated transcription of C/EBPβ, which is essential for the early stage of adipocyte differentiation. Using dual-luciferase assay, we validate that knockdown of FTO reduces expression of JAK2 in an m6A-dependent manner. Furthermore, we find that m6A "reader" protein YTHDF2 directly targets m6A-modified transcripts of JAK2 and accelerates mRNA decay, which results in decreased JAK2 expression and inactivated JAK2-STAT3-C/EBPβ signaling, thereby inhibiting adipogenesis. Collectively, our results provide a novel insight into the molecular mechanism of m6A methylation in post-transcriptional regulation of JAK2-STAT3-C/EBPβ signaling axis and highlight the crucial role of m6A modification and its modulators in adipogenesis.