跳转至内容
Merck
  • Resveratrol Ameliorates Lipid Droplet Accumulation in Liver Through a SIRT1/ ATF6-Dependent Mechanism.
Resveratrol Ameliorates Lipid Droplet Accumulation in Liver Through a SIRT1/ ATF6-Dependent Mechanism.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-12-12)
Rui Zhou, Long Yi, Xikun Ye, Xianglong Zeng, Kai Liu, Yu Qin, Qianyong Zhang, Mantian Mi
摘要
Lipid droplets (LDs) are dynamic organelles that store neutral lipids during times of energy excess, and an increased accumulation of LDs in the liver is closely linked to hepatic steatosis. Our previous studies suggested that resveratrol (RSV) supplement could improve hepatic steatosis, but the underlying mechanism, particularly which related to LD accumulation, has not yet been elucidated. A high-fat diet (HFD) and palmitic acid were used to induce hepatic steatosis in mouse liver and hepatocytes, respectively. The effects of RSV on LD accumulation were analyzed in vivo and in vitro. The effects of RSV on the expression levels of LD-associated genes (ATF6, Fsp27β/CIDEC, CREBH, and PLIN1) were measured by qRT-PCR and western blot assays, followed by KD or overexpression of SIRT1 and ATF6 with small interfering RNAs or overexpressed plasmids, respectively. The dual luciferase reporter assay, chromatin immunoprecipitation assay, coimmunoprecipitation, and proximity ligation assay were utilized to clarify the mechanism of transcriptional regulation and possible interaction between SIRT1 and ATF6. There was a significant increase in the accumulation of LDs in liver and hepatocytes during the process of HFD-induced steatosis, respectively, which was significantly inhibited by RSV supplementation. RSV notably activated SIRT1 expression and decreased the expression levels of ATF6, Fsp27β/CIDEC, CREBH, and PLIN1, which are associated with LD accumulation. Interestingly, the inhibitory effects of RSV on LD accumulation and the associated expression of genes in hepatocytes were abrogated or strengthened with SIRT1 silencing or overexpression, respectively. On the contrary, the benefits of RSV in hepatocytes were eliminated or aggravated when transfected with the overexpressed ATF6 or ATF6 siRNA, respectively. Furthermore, we found that RSV stimulated SIRT1 expression significantly, which was followed by increased deacetylation and inactivation of ATF6, resulting in a positive feedback loop for SIRT1 transcription associated with ATF6 binding to the SIRT1 promoter region. Taken together, these findings indicate that RSV supplementation improves hepatic steatosis by ameliorating the accumulation of LDs, and this might be partially mediated by a SIRT1/ATF6-dependent mechanism.