跳转至内容
Merck
  • Characteristics of the interactions between acid glycosaminoglycans and proteins in normal human plasma as revealed by the behaviour of the protein-polysaccharide complexes in ultrafiltration and chromatographic procedures.
Characteristics of the interactions between acid glycosaminoglycans and proteins in normal human plasma as revealed by the behaviour of the protein-polysaccharide complexes in ultrafiltration and chromatographic procedures.
Clinica chimica acta; international journal of clinical chemistry (1992-03-31)
A Calatroni, R Vinci, A M Ferlazzo
摘要
Acid glycosaminoglycans were isolated from normal human plasma: (a) following fractionation of plasma (with protease inhibitors) on Sephadex G-200; (b) by ultrafiltration through membranes with retention of molecules above 50 kDa, with and without previous addition of NaCl, Triton X-100, urea, or guanidine HCl; (c) by filtering on Ecteola-cellulose either untreated plasma or after treatment with NaCl, urea, Triton X-100, papain or NaOH. More than 95% of plasma glycosaminoglycans interact with plasma proteins to give complexes that exhibit reproducible behaviour on Sephadex G-200 and are retained by ultrafiltration membranes, which the 12-20 kDa polysaccharide chains do filter. High charge plasma glycosaminoglycans show ionic interactions with proteins, while low charge glycosaminoglycan interactions are resistant to Ecteola charged groups, to 0.5% Triton and 4 M urea, while not to 4 M guanidine HC1. Some glycosaminoglycan-protein complexes appear resistant to proteolysis, suggesting that they may originate from lymphocytes. The simple method utilized for plasma GAG measurement may represent an useful tool in clinical practice.
材料
货号
品牌
产品描述
Sigma-Aldrich
Ecteola Cellulose, coarse