跳转至内容
Merck
  • Context-enriched interactome powered by proteomics helps the identification of novel regulators of macrophage activation.
Context-enriched interactome powered by proteomics helps the identification of novel regulators of macrophage activation.
eLife (2018-10-12)
Arda Halu, Jian-Guo Wang, Hiroshi Iwata, Alexander Mojcher, Ana Luisa Abib, Sasha A Singh, Masanori Aikawa, Amitabh Sharma
摘要
The role of pro-inflammatory macrophage activation in cardiovascular disease (CVD) is a complex one amenable to network approaches. While an indispensible tool for elucidating the molecular underpinnings of complex diseases including CVD, the interactome is limited in its utility as it is not specific to any cell type, experimental condition or disease state. We introduced context-specificity to the interactome by combining it with co-abundance networks derived from unbiased proteomics measurements from activated macrophage-like cells. Each macrophage phenotype contributed to certain regions of the interactome. Using a network proximity-based prioritization method on the combined network, we predicted potential regulators of macrophage activation. Prediction performance significantly increased with the addition of co-abundance edges, and the prioritized candidates captured inflammation, immunity and CVD signatures. Integrating the novel network topology with transcriptomics and proteomics revealed top candidate drivers of inflammation. In vitro loss-of-function experiments demonstrated the regulatory role of these proteins in pro-inflammatory signaling.
材料
货号
品牌
产品描述
Sigma-Aldrich
佛波醇12-十四酸酯13-乙酸酯, ≥99% (TLC), film or powder
Sigma-Aldrich
MISSION® esiRNA, targeting human GBP1