返回
ROTOR FOR EPPENDORF
®
5810/5810R CENTRIFUGES
Products
产品分类
rotors (3)
制造商
包装大小
筛选产品
Showing 1-3 of 3
比较
货号
说明
价格
EP5820710004
ROTOR FOR EPPENDORF
®
5810/5810R CENTRIFUGES,
A-2-DWP-AT, aerosol-tight, for 5810/5810R, with 2 buckets, 2 caps & 2 plate carriers
展开
查看定价
RA481750-EP
ROTOR FOR EPPENDORF
®
5810/5810R CENTRIFUGES,
A-4-81, includes 7x50 mL conical buckets
展开
查看定价
RA2DWPAT-EP
ROTOR FOR EPPENDORF
®
5810/5810R CENTRIFUGES,
A-2-DWP-AT, aerosol-tight