返回
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB
别名:
Milligard
®
0.2 µm标称,Opticap
®
XL囊式
Products
产品分类
capsule filters (12)
品牌
Millipore (12)
灭菌适用性
机构
长度(cm)
长度(in)
质量水平
疏水性
外壳材料
膜材料
支架材料
孔径(µm)
无菌性
筛选产品
Showing 1-12 of 12
比较
货号
说明
价格
KW03A04HH3
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
inlet connection diam. 9/16 in., Opticap
®
XL 4, pore size 0.2 μm, cartridge nominal length 4 in. (10 cm)
展开
查看定价
KW03A02FF3
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
Opticap
®
XL 2, inlet connection diam. 3/4 in., pore size 0.2 μm, cartridge nominal length 2 in. (5 cm)
展开
查看定价
KW03A02HH3
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
Opticap
®
XL 2, inlet connection diam. 9/16 in., pore size 0.2 μm, cartridge nominal length 2 in. (5 cm)
展开
查看定价
KW03A04FF3
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
Opticap
®
XL 4, inlet connection diam. 3/4 in., pore size 0.2 μm, cartridge nominal length 4 in. (10 cm)
展开
查看定价
KW03A04TT3
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
inlet connection diam. 1.5 in., Opticap
®
XL 4, pore size 0.2 μm, cartridge nominal length 4 in. (10 cm)
展开
查看定价
KW03A10TT1
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
inlet connection diam. 1.5 in., Opticap
®
XL 10, pore size 0.2 μm, cartridge nominal length 10 in. (25 cm)
展开
查看定价
KW03A02TT3
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
Opticap
®
XL 2, inlet connection diam. 1.5 in., pore size 0.2 μm, cartridge nominal length 2 in. (5 cm)
展开
查看定价
KW03A05HH1
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
inlet connection diam. 9/16 in., Opticap
®
XL 5, pore size 0.2 μm, cartridge nominal length 5 in. (12.5 cm)
展开
查看定价
KW03A10HH1
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
inlet connection diam. 9/16 in., Opticap
®
XL 10, pore size 0.2 μm, cartridge nominal length 10 in. (25 cm)
展开
查看定价
KW03A05FF1
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
inlet connection diam. 3/4 in., Opticap
®
XL 5, pore size 0.2 μm, cartridge nominal length 5 in. (12.5 cm)
展开
查看定价
KW03A05TT1
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
inlet connection diam. 1.5 in., Opticap
®
XL 5, pore size 0.2 μm, cartridge nominal length 5 in. (12.5 cm)
展开
查看定价
KW03A10FF1
Opticap XL4 Milligard
0.2 µm 9/16 in.HB/HB,
inlet connection diam. 3/4 in., Opticap
®
XL 10, pore size 0.2 μm, cartridge nominal length 10 in. (25 cm)