返回
MAGic Clamp
™
magnetic clamp
Products
产品分类
clamps (9)
制造商
筛选产品
Showing 1-9 of 9
比较
货号
说明
价格
Z742439
MAGic Clamp
™
magnetic clamp,
adjustable flask clamp holds 125 mL, 250 mL, or 500 mL flasks
展开
查看定价
Z763500
MAGic Clamp
™
magnetic clamp,
holds 250 mL Erlenmeyer flasks (max. 8)
展开
查看定价
Z763497
MAGic Clamp
™
magnetic clamp,
holds 125 mL Erlenmeyer flasks (max. 12)
展开
查看定价
Z742438
MAGic Clamp
™
magnetic clamp,
holds 1 microplate
展开
查看定价
Z763527
MAGic Clamp
™
magnetic clamp,
holds 1000 mL Erlenmeyer flasks (max. 4)
展开
查看定价
Z763535
MAGic Clamp
™
magnetic clamp,
holds 2000 mL Erlenmeyer flasks (max. 2)
展开
查看定价
Z763519
MAGic Clamp
™
magnetic clamp,
holds 500 mL Erlenmeyer flasks (max. 5)
展开
查看定价
Z742448
MAGic Clamp
™
magnetic clamp,
holds 3000 mL Erlenmeyer flask
展开
查看定价
Z763489
MAGic Clamp
™
magnetic clamp,
holds 50 mL Erlenmeyer flasks (max. 20)