返回
Kern PFB series precision balances
Products
产品分类
balances (7)
制造商
直径(mm)
最大称重量(g)
包装大小
筛选产品
Showing 1-7 of 7
比较
货号
说明
价格
Z742847
Kern PFB series precision balances,
Kern PFB 1200-2, weighing capacity 1200 g, accuracy: 0.01 g, precision: ±0.03 g
展开
查看定价
Z742849
Kern PFB series precision balances,
Kern PFB 2000-2, weighing capacity 2000 g, accuracy: 0.01 g, precision: ±0.03 g
展开
查看定价
Z742844
Kern PFB series precision balances,
Kern PFB 120-3, weighing capacity 120 g, accuracy: 0.001 g, precision: ±0.003 g
展开
查看定价
Z742850
Kern PFB series precision balances,
Kern PFB 3000-2, weighing capacity 3000 g, accuracy: 0.01 g, precision: ±0.03 g
展开
查看定价
Z742845
Kern PFB series precision balances,
Kern PFB 200-3, weighing capacity 200 g, accuracy: 0.001 g, precision: ±0.003 g
展开
查看定价
Z742846
Kern PFB series precision balances,
Kern PFB 300-3, weighing capacity 300 g, accuracy: 0.001 g, precision: ±0.003 g
展开
查看定价
Z742852
Kern PFB series precision balances,
Kern PFB 6000-1, weighing capacity 6000 g, accuracy: 0.1 g, precision: ±0.3 g