返回
External Solenoid valve
Products
产品分类
valves (2)
品牌
Milli-Q (2)
相容性
筛选产品
Showing 1-2 of 2
比较
货号
说明
价格
EXTSV00A1
External Solenoid valve,
Regulates feed water flow, for use with Milli-Q
®
IQ/IX systems
展开
查看定价
ZLXL00ESV
External Solenoid valve,
For use with Milli-Q
®
CLX / HR / HX systems