返回
ech
2
o
®
紫外线灯
Products
产品分类
UV lamps (4)
品牌
Milli-Q (4)
相容性
质量水平
筛选产品
Showing 1-4 of 4
比较
货号
说明
价格
ZFA10UVA1
ech
2
o
®
紫外线灯,
Component of the A10
®
TOC monitor, for use with Milli-Q
®
IQ Systems
展开
查看定价
ZIQUVLPA1
ech
2
o
®
紫外线灯,
for use with Milli-Q
®
IQ Systems, Mercury-free UV lamp for photooxidation of organic compounds
展开
查看定价
ZIXUVLPA1
ech
2
o
®
紫外线灯,
Protects stored water against biofilm buildup in Milli-Q
®
IX & IQ 7003/05/10/15 storage tanks
展开
查看定价
IQ7XUVKT1
ech
2
o
®
紫外线灯,
Protects stored water against biofilm buildup in Milli-Q
®
IQ storage tanks