返回
Ace Enlarging Adapter
Products
产品分类
adapters (4)
制造商
筛选产品
Showing 1-4 of 4
比较
货号
说明
价格
Z158232
Ace Enlarging Adapter,
female joint: ST/NS 29/42, male joint: ST/NS 24/40
展开
查看定价
Z158291
Ace Enlarging Adapter,
female joint: SJ 35/20, male joint: ST/NS 24/40
展开
查看定价
Z158305
Ace Enlarging Adapter,
female joint: SJ 35/25, male joint: ST/NS 24/40
展开
查看定价
Z158267
Ace Enlarging Adapter,
female joint: ST/NS 45/50, male joint: ST/NS 29/42