Merck
流体运输与储存
Mobius®一次性储存和运输系统,包括方便移动的带轮子桶状容器、用于规整储液的多层袋子架、用于模组化液体生产管理的箱形容器。该系列产品均带有Mobius®标志。
产品直达:
Mobius®储存系统可为您的一次性生产提供高效、符合人体工程学且可扩大的流体管理解决方案。我们可为产品储存、转移、搬运和运输提供各种选择,包括聚乙烯(PE)药桶、不锈钢废料桶、可拆卸废料桶和容器推车。   
优势包括:
  • 操作失误减少
  • 工艺效率提高
  • 更高的灵活性和移动性
  • 支持一次性技术
相关资源
  • Tech Brief: Holistic Buffer Preparation
    Buffer preparation is a critical but often undervalued aspect of biologics production. It involves a significant volume of buffers, with 15 different solutions used on average per process. This document discusses the optimization of in-house buffer preparation and potential considerations for outsourcing, highlighting the potential for a combined approach.
  • Tech Brief: Shipping Qualification of Mobius® Large Liquid Transportation
    Safe and efficient transportation of large bulk liquid substances, including critical items like sterile water, buffers, and drug products, is essential. We completed an extensive ISTA shipping validation to ensure the safest transportation. When considering transportation for these substances, various factors need to be evaluated for peace of mind.
  • Brochure: Single-use Fluid Management: 2D and 3D Assemblies, Filtration, Storage, and Transportation Solutions
    Mobius® offers 2D and 3D assemblies, filtration, storage, and transportation solutions for fluid management. These solutions include various container options and are designed to reduce errors, minimize risks, and improve efficiency. They provide flexibility, mobility, and support for single-use technologies at different certification levels.
  • Tool: mAbsizer™ Calculator
    mAb Expression, Purification, and Formulation
聚乙烯(PE)药桶和废料桶
我们的聚乙烯药桶适于培养基和缓冲液的工艺级一次性生产应用。这些可堆叠的药桶具有50L、100L和200L的规格选择,可容纳带有顶部和底部容器接口的一次性标准组件,如在无菌应用中连接管组的过滤器囊。可与手推车配合使用,搬运方便。
可拆卸废料桶
可为缓冲液制备、培养基输送和成品配制等生产阶段提供简单方便的储存,可安全地容纳从100L到1000L的工艺容器。闲置时,可拆卸和堆叠起来,提高拥挤洁净室的空间利用率。
不锈钢桶
Mobius® 不锈钢桶可在生物制药中为培养基、缓冲液和附加中间体的使用和储存提供先进的解决方案。标准规格从200L到1500L不等,可容纳Mobius® 和其他制造商生产的各种工艺容器。
  • 单一大开口,便于容纳废料和容器定位。
  • 304/304L不锈钢结构
  • 便捷的滑动托盘
  • 简单的折叠设计,灌装时无需操作员或机械协助
  • 在一次性构建中的不含动物源性组分材料
  • 可容纳定制设计的2,000L至3,000L的不锈钢桶
容器推车
Mobius®容器推车使2D袋的搬运和储存变得简单而安全,同时还能保证容器及其内部物质的完整性。移动方便,占地面积小,可提高运输灵活性。每辆推车可装载高达6件规格为10 L、20 L和50 L的工艺容器。推车含一筐六篮,容器篮可向前滑动并固定位置,以便于装卸。还有一个便捷把手,方便操作员倾斜推车,倾倒流体。