主要文件
Z379409
Hanna Piccolo® pH meter
Piccolo
登录查看公司和协议定价
About This Item
UNSPSC代码:
41115603
价格与库存信息目前不能提供
描述
temperature compensation 0 - 70 °C (automatic with integral temperature sensor)
特点
calibration manual dual point, ±1 pH
制造商/商品名称
HI98111
pH值测量范围
0-14, accuracy: 0.01, resolution: 0.01
电极长度
90 mm
重量
400 gincluding calibration screwdriver, ABS carrying case, and batteries
70 g
电池电量
3 × 1.4 V button cell, 100 hr life (continuous operation)
正在寻找类似产品? 访问 产品对比指南
一般描述
Hand-held stick-style pH/temperature meters with rugged design for field use. "Amplified electrode" delivers strong signal to the meter, eliminating the vulnerability to noise, humidity, and dirt that is characteristic of conventional meters.
法律信息
Piccolo is a registered trademark of Hanna Instruments
历史批次信息供参考:
分析证书(COA)