Iupac Name
sodium ((2R,3S,4R,5R)-5-(2-amino-6-hydroxy-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate
Inchi Code
1S/C10H16N5O14P3.2Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);;/q;2*+1/p-2/t3-,5-,6-,9-;;/m1../s1
InChI key
FIZIYLKEXVIRHJ-LGVAUZIVSA-L
储存温度
Inert atmosphere,Store in freezer, under -20C
Purity
89%
Country of Origin
CN
Shipping Temperature
Shipped in cold pack
Physical Form
solid
Product Link
https://www.ambeed.com/products/56001-37-7.html
Synonym
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-1H-purin-9(6H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt
Signal Word
Warning
Hazard Statements
Precautionary statements
我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.
联系客户支持