Ampule Opener from Bel-Art
Product No. Z693847
The innovative box-style design enables one-handed opening of up to 3 ampules simultaneously by pressing down on the lid. The ampule necks snap cleanly and are contained within the device, keeping fingers clear of sharp edges.
After emptying the ampules, invert the device over a waste receptacle to safely dispose of both the ampules and their tops.
Network error: Failed to fetch