Eosin B (bluish) (C.I.45400)
Synonym(s):
Eosin B (bluish) (C.I.45400)
Empirical Formula (Hill Notation):
C20H6Br2N2Na2O9
CAS Number:
Molecular Weight:
624.06
Products
Brand
Formula Weight
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing