V900118
Calcium acetate monohydrate
Vetec™, reagent grade, 98%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
Ca(CH3CO2)2 · H2O
CAS Number:
Molecular Weight:
176.18
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
98%
grade:
reagent grade
form:
powder
grade
reagent grade
product line
Vetec™
Assay
98%
form
powder
SMILES string
[H]O[H].CC(=O)O[Ca]OC(C)=O
InChI
1S/2C2H4O2.Ca.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2
InChI key
XQKKWWCELHKGKB-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dilek Teker et al.
Materials science & engineering. C, Materials for biological applications, 48, 579-585 (2015-01-13)
An innovative multi-layer coating comprising a bioactive compound layer (consisting of hydroxyapatite and calcium titanate) with an underlying titanium oxide layer (in the form of anatase and rutile) has been developed on Grade 4 quality commercially pure titanium via a
Hamid Esfahani et al.
Journal of colloid and interface science, 443, 143-152 (2014-12-30)
Surface modification of electrospun polymeric membrane surfaces is a critical step towards the separation process including protein adsorption. In this study, the electrospun Nylon fibers was incorporated with positively charged zinc doped hydroxyapatite (HAp) nanoparticles to study the adsorption of
S Yoon et al.
Cell death & disease, 5, e1494-e1494 (2014-10-31)
Apoptosis, which is anti-inflammatory, and necrosis, which is pro-inflammatory, represent the extremes of the cell death spectrum. Cell death is complex and both apoptosis and necrosis can be observed in the same cells or tissues. Here, we introduce a novel
Tanja Rozman Peterka et al.
Journal of pharmaceutical and biomedical analysis, 110, 67-75 (2015-03-26)
Tacrolimus is macrolide drug that is widely used as a potent immunosuppressant. In the present work compatibility testing was conducted on physical mixtures of tacrolimus with excipients and on compatibility mixtures prepared by the simulation of manufacturing process used for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service