Skip to Content
Merck
All Photos(1)

Key Documents

1643907

USP

Terfenadine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Terfenadone, 1-[4-(1,1-Dimethylethyl)phenyl]-4-[4-(hydroxydiphenylmethyl)-1-piperidinyl]-1-butanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H39NO2
CAS Number:
Molecular Weight:
469.66
Beilstein:
6352257
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4

InChI

1S/C32H39NO2/c1-31(2,3)26-18-16-25(17-19-26)30(34)15-10-22-33-23-20-29(21-24-33)32(35,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29,35H,10,15,20-24H2,1-3H3

InChI key

YCADFOXVGUGHRV-UHFFFAOYSA-N

General description

Terfenadine Related Compound A also called as terfenadine is derivative of terfenadine. It is mostly used as starting point for the designing of high-affinity inhibitors of CYP2J2.

Application

It was used as internal standard during determination of terfenadine in human plasma using HPLC-MS/MS analysis.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective inhibitors of CYP2J2 related to terfenadine exhibit strong activity against human cancers in vitro and in vivo.
Chen C
Journal of Pharmacology and Experimental Therapeutics, 329(3), 908-918 (2009)
High sensitivity high performance liquid chromatography electrospray tandem mass spectrometry determination of terfenadine in human plasma.
Lau, Yau Yi, Paul H. Anderson, and Rasmy Talaat.
Journal of Liquid Chromatography and Related Technologies, 19.16, 2669-2679 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service