Skip to Content
Merck
All Photos(1)

Documents

1044470

USP

Atomoxetine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

α-[2-(Methylamino)ethyl]benzyl alcohol, 3-(Methylamino)-1-phenylpropan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH2CH2NHCH3)OH
CAS Number:
Molecular Weight:
165.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

atomoxetine

manufacturer/tradename

USP

mp

59-64 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CNCCC(O)c1ccccc1

InChI

1S/C10H15NO/c1-11-8-7-10(12)9-5-3-2-4-6-9/h2-6,10-12H,7-8H2,1H3

InChI key

XXSDCGNHLFVSET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atomoxetine Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Atomoxetine Hydrochloride

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service