Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5885124

Sulfuric acid

95-99%, univAR

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352322

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

product line

univAR

Assay

95-99%

packaging

glass bottle of 1 × 2.5 L (SAAR5885124LCA)
poly bottle of 1 × 2.5 L (SAAR5885124LP)
poly bottle of 1 × 5 L (SAAR5885124LL)

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service