Skip to Content
Merck
All Photos(1)

Documents

PHL82683

Withanone

phyproof® Reference Substance

Synonym(s):

(22R)-6α,7α-Epoxy-5,17,22-trihydroxy-1-oxo-5α-ergosta-2,24-dien-26-oic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H38O6
CAS Number:
Molecular Weight:
470.60
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

O=C1[C@@]2(C)[C@]([C@@H](O3)[C@@H]3[C@]4([H])[C@]2([H])CC[C@@]5(C)[C@@]4([H])CC[C@@]5([C@H](C)[C@]6([H])OC(C(C)=C(C)C6)=O)O)(O)CC=C1

InChI

1S/C28H38O6/c1-14-13-19(33-24(30)15(14)2)16(3)27(31)12-9-17-21-18(8-11-25(17,27)4)26(5)20(29)7-6-10-28(26,32)23-22(21)34-23/h6-7,16-19,21-23,31-32H,8-13H2,1-5H3/t16-,17+,18+,19-,21+,22+,23+,25+,26+,27+,28+/m1/s1

InChI key

FAZIYUIDUNHZRG-PCTWTJKKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Anticancer Agent: Withanone has been recognized as an emerging anticancer agent. It exhibits significant potential in combating cancer through molecular mechanisms that have been validated by both experimental and computational evidence, emphasizing its role in therapeutic applications (Stephen et al., 2024).

  • Phytotherapeutic Potential: Withanone, derived from Withania somnifera, is highlighted for its broad phytotherapeutic potential. It impacts various health conditions positively, particularly through molecular mechanisms that enhance bodily functions and overall health (Yadav et al., 2024).

  • Salinity Stress Response in Plants: Research has demonstrated that withanone can modulate physiological and biochemical traits in plants, enhancing their response to salinity stress. This adaptation includes the modulation of withanolide content, which has implications for agricultural practices and crop resilience (Thorat et al., 2023).

  • Epigenetic Cancer Therapy: Withanone is studied for its effects on DNA methyltransferases, suggesting its potential use in epigenetic cancer therapy. Its ability to modulate epigenetic markers opens new avenues for cancer treatment strategies (Kumar et al., 2023).

  • Pharmacokinetics in Neuroprotection: The pharmacokinetics of withanone have been analyzed to determine its distribution in rat plasma and brain, providing foundational data that supports its use as a neuroprotective agent. This research is crucial for developing therapeutic strategies for neurological disorders (Dadge et al., 2023).

Other Notes

This compound is commonly found in plants of the genus: withania

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

420.8 °F

Flash Point(C)

216 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service