Skip to Content
Merck
All Photos(1)

Key Documents

G005962

HPLC Separation of an Oligonucleotide and Its Degradation Fragments on Proteomix® SAX-NP5

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777228)
mobile phase: (A) 25 mM Tris, 1 mM EDTA 10% acetonitrile (8.0 pH); (B) A + 1.0 M NaCl
gradient: 0 to 75% B in 50 min
flow rate: 0.5 mL/min
detector:
sample: Oligonucleotide (MW 12196) and its degradation fragments

suitability

application for HPLC

application(s)

forensics and toxicology
life science and biopharma

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service