Skip to Content
Merck
All Photos(1)

Documents

96141

Supelco

4-Amino-3-nitrophenol

analytical standard

Synonym(s):

4-Hydroxy-2-nitroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(NO2)OH
CAS Number:
Molecular Weight:
154.12
Beilstein:
2210196
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Assay

98.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

150-154 °C (lit.)

format

neat

SMILES string

Nc1ccc(O)cc1[N+]([O-])=O

InChI

1S/C6H6N2O3/c7-5-2-1-4(9)3-6(5)8(10)11/h1-3,9H,7H2

InChI key

IQXUIDYRTHQTET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service