Skip to Content
Merck
All Photos(1)

Documents

821450

TOYOPEARL® Butyl-600M Bulk Media

bottle of 1 L, 65 μm particle size

Synonym(s):

TOYOPEARL® Butyl-600M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

description

Adsorption capacity (50 +/- 10 mg/mL (Lysozyme))
Clean in place (CIP) (0.5 M NaOH or 0.1 M HCl)
Formulation (72% (v/v) slurry in 20% (v/v) ethanol)
Phase filter (Butyl)
Shipping solvent (20% (v/v) ethanol)

product line

TOYOPEARL®

feature

Molecular weight range (Excl. limit: 1,000,000 Da)

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 21450

parameter

≤50% max. pressure (organic solvent)
≤50% organic solvent
3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

C4 (butyl) phase

particle size

65 μm

pore size

750 Å

pH range

1-13

capacity

50 g/L±10 adsorption capacity (lysozyme)

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl Butyl-600 resin has the second highest hydrophobicity of the HIC ligands offered by Tosoh Bioscience. Its pore size is between that of Toyopearl Butyl-650 and Toyopearl SuperButyl-550 media. It has been optimized for the purification of monoclonal antibodies and is typically used as an intermediate purification or polishing step.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service