64753-U
Specialty Glass Column
glass coarse frit, PTFE stopcock, column L × O.D. × I.D. 400 mm × 22 mm × 19 mm, 515mm overall
Synonym(s):
LC COL 400MM X 19MMID W/FRIT STOPCOCK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
PTFE stopcock
glass coarse frit
technique(s)
LPLC: suitable
column L × O.D. × I.D.
400 mm × 22 mm × 19 mm, 515mm overall
compatibility
for use with US EPA Protocol
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
Described in US EPA Protocol - Fill these columns with appropriate adsorbents when cleaning or drying environmental samples for pesticide or priority pollutant analyses.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service