Documents
57692
RheFlex® PEEK Fittings
Ferrule pack, 1/16 in. tubing
About This Item
UNSPSC Code:
41115700
packaging
pkg of 5 ea
compatibility
1/16 in. tubing
General description
PEEK (polyetheretherketone) fingertight fittings (ferrule pack) are convenient, inert, and biocompatible. Use these fittings with 1/16 in. O.D. PEEK, stainless steel, titanium, Tefzel, or PTFE tubing. PEEK fittings are compatible with all HPLC solvents (avoid concentrated sulfuric and nitric acids), and can be used at temperatures up to 150° C. Unlike stainless steel ferrules, PEEK ferrules do not permanently lock into place on the tubing. This allows the interchanging of fittings with tubing and columns, from manufacturer to manufacturer (e.g. from Agilent to Waters), and still form a zero dead volume, leak-free connection.
Features and Benefits
For Rheodyne valves. Hold to 5000 psi (350 kg/cm²).
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
RheFlex is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)