502413
Upchurch PEEK Microtight® Connector
Butt connects 2 pieces of capillary. True zero dead volume design, pkg of 2 ea
Synonym(s):
Microtight® Union
All Photos(1)
About This Item
UNSPSC Code:
41115700
Recommended Products
description
zero dead volume
packaging
pkg of 2 ea
General description
Connects 2 pieces of capillary tubing. True zero-dead volume design -- tubing pieces butt together. Use gauge plug (included) to properly seat tubing. Includes 2 MicroFingertight fittings.
Legal Information
Microtight is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)