48852-U
EPA Purgeable B
analytical standard, 200 μg/mL each component in methanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
77101502
grade
analytical standard
Agency
EPA 624
description
Separate Source
feature
standard type calibration
packaging
ampule of 1 × 1 mL
concentration
200 μg/mL each component in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-8°C
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Analyte
Description
Benzene
Bromodichloromethane
Bromoform
1,2-Dichloroethane
trans-1,2-Dichloroethylene
cis-1,3-Dichloropropene
trans-1,3-Dichloropropene
Ethylbenzene
1,1,2,2-Tetrachloroethane
Toluene
1,1,1-Trichloroethane
See All (11)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT SE 1
Target Organs
Eyes,Central nervous system
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service