Skip to Content
Merck
All Photos(1)

Documents

47351

Supelco

EPA 610-N PAH Kit

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
eCl@ss:
32161000

grade

analytical standard

Agency

EPA 610

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

pkg of 1 kit

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

General description

EPA 610-N PAH Kit can be utilized for identifying and quantifying polynuclear aromatic hydrocarbons (PAHs) in municipal and industrial wastewater. The pre-requisite of EPA 610 method is the use of gas chromatography with a selective detector or for gas chromatography/mass spectrometry. The analyst may be better equipped to choose between GC and HPLC. Packed GC columns are described in the 610 method, but chromatographic conditions can be changed (e.g., capillary columns may be used) if the changes do not make the method "less accurate or less precise" than the standard method.
Essentially, our EPA 610-N PAH kit contains 16 individually packaged polynuclear aromatic hydrocarbon neats for customizing solutions to meet the unique needs of the laboratory. Each PAH has been tested by GC for purity.

Application

EPA 610-N PAH Kit may be used as an anaytical reference standard mix for the quantification and separation of the polycyclic aromatic hydrocarbons in mixtures using liquid chromatography coupled to electrochemical detection with mass spectrometry (LC-EC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Kit Components Only

Product No.
Description

  • Acenaphthylene .1 g

  • Anthracene 5 g

  • Benzo[a]anthracene .1 g

  • Benzo[a]pyrene .1 g

  • Benzo[b]fluoranthene .05 g

  • Benzo[ghi]perylene .025 g

  • Benzo[k]fluoranthene .05 g

  • Chrysene(93%) .1 g

  • Dibenzo[a,h]anthracene .1 g

  • Fluoranthene 5 g

  • Fluorene 5 g

  • Indenol[1,2,3-cd]pyrene .01 g

  • Naphthalene 5 g

  • Phenanthrene 5 g

  • Pyrene 5 g

  • Acenaphthene 5 g

See All (16)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liquid chromatography-electrochemistry-mass spectrometry of polycyclic aromatic hydrocarbons
MS, et al.
Analytical and Bioanalytical Chemistry, 378(4), 917-925 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service