27002-U
Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100
volume 7 mL, amber glass vial, O.D. × H 17 mm × 60 mm, thread for 15-425
Synonym(s):
PK100 7ML AMBER VIAL,SCREW TOP, SOLID CAP W/PTFE LINER
About This Item
Recommended Products
material
F217/PTFE cap liner
PTFE liner
amber glass vial
green melamine resin cap (solid-top)
feature
closure type green melamine solid-top cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
17 mm × 60 mm
volume
7 mL
fitting
thread for 15-425
Looking for similar products? Visit Product Comparison Guide
Related Categories
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service