Documents
26705
Hamilton® chaney adapter
for use with Hamilton 800/1800 series (5-500 μL)
About This Item
UNSPSC Code:
41122001
material
aluminum
feature
For consistent, reproducible injections
Prevents syringe plunger bending
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 32146
compatibility
for use with Hamilton 800/1800 series (5-500 μL)
General description
Recommended for sample reproducibility.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)