Skip to Content
Merck
All Photos(2)

Key Documents

18514

Sigma-Aldrich

Kieselguhr

greener alternative

calcined, purified

Synonym(s):

Infusorial earth

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

form

powder

quality

calcined, purified

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

impurities

≤1% HCl solubles

loss

≤1% loss on ignition

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Fe: ≤500 mg/kg

greener alternative category

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kieselguhr (diatomite, infusorial earth) is the fossilized skeletons of diatom algae. It is mainly made up of silica with many unique features like high porosity, small particle size, low density and thermal conductivity. It is primarily utilized as a source to manufacture filter aids, fire-bricks and adsorbents. Kieselguhr is the main component of the Berkefeld filter used for purifying water. It is also utilized in the filtration process of beer brewing.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Find details here.

Application

Kieselguhr (infusorial earth) may be used in the following processes:
  • Estimation of total fats.
  • As a catalyst in the synthesis of ketones and aldehydes via oxidation of corresponding oximes with manganese dioxide.
  • Green synthesis of β-nitroalkenes via Michael addition of 1,3-dicarbonyl compounds, by a grinding method in the absence of solvent and catalyst.
  • Preparation of KMnO4-kieselguhr reagent for use in the oxidation of primary alcohols to aldehydes with shorter reaction time.
Used for greener amide bond formation from amines and carboxylic acids after thermal activation at high temperature.

Clean, reusable and low cost heterogeneous catalyst for amide synthesis

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Household Water Treatment following Emergencies and Disasters- A Review
Gambhir RS, et al.
Indian Journal of Public Health Research & Development, 4(3), 249-252 (2013)
Manufacture of ceramic bricks using recycled brewing spent Kieselguhr.
Ferraz E, et al.
Materials and Manufacturing Processes, 26(10), 1319-1329 (2011)
Removal of uranium from aqueous solutions by diatomite (Kieselguhr).
Olmez AS, et al.
J. Radioanal. Nucl. Chem., 240(3), 973-976 (1999)
Catalyst-free and solvent-free Michael addition of 1,3-dicarbonyl compounds to nitroalkenes by a grinding method.
Xie ZB, et al.
Beilstein Journal of Organic Chemistry, 8(1), 534-538 (2012)
Sudhakara Gujjala et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 215-223 (2016-09-23)
The objective of the present study was to investigate the preventive effects of hydro-alcoholic extract of Caralluma fimbriata (CFE) and Metformin (Met) against high-fat diet (HF-diet) induced alterations in lipid metabolism in Wistar rats. The experimental animals were divided into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service