Skip to Content
Merck
All Photos(1)

Key Documents

11965

Supelco

Coated GC Packing

phase 10% SP-1200 + 1% H3PO4, matrix 80/100 Chromosorb W AW support, bottle of 20 g

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

packaging

bottle of 20 g

parameter

25-200 °C temp. range

matrix

80/100 Chromosorb W AW support

matrix active group

10% SP-1200 + 1% H3PO4 phase

compatibility

for analyte group fatty acids

General description

Ready-to-use coated packing for those wanting to pack their own columns.

Application

C2-C5 volatile fatty acids

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service