Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2125

Sigma-Aldrich

Anti-JAM-A Antibody, clone 2E9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Junctional adhesion molecule A;Junctional adhesion molecule 1, JAM-1, Platelet F11 receptor, Platelet adhesion molecule 1, PAM-1, CD321

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E9, recombinant monoclonal

description

2E9 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 32.58 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... F11R(50848)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2E9 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Junctional adhesion molecule A (JAM-A). It targets an epitope within 19 amino acids from the N-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal, extracellular domain of human Junctional adhesion molecule A (JAM-A).

Application

Quality Control Testing

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected JAM-A in MCF-7 cell lysate.

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected JAM-A in CaCo2 cell lysate and 1:1,000 dilution in HepG2 and HeLa cell lysates.

Affinity Binding Assay:: A representative lot of this antibody bound JAM-A peptide with a KD of 1.2 x 10-6 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected JAM-A in human liver tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected JAM-A in MCF-7 cell lysate.

Target description

Junctional adhesion molecule A (UniProt: Q9Y624; also known as JAM-A, Junctional adhesion molecule 1, JAM-1, Platelet F11 receptor, Platelet adhesion molecule 1, PAM-1, CD321) is encoded by the F11R (also known as JAM1, JCAM) gene (Gene ID: 50848) in human. JAM-A is a single-pass type I membrane protein that is localized at tight junctions of both epithelial and endothelial cells. It is also expressed in circulating platelets and leukocytes. It is synthesized with a signal peptide (aa 1-27), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 28-238), a transmembrane domain (aa 239-259), and a cytoplasmic domain (aa 260-299). The extracellular domain contains a membrane distal VH-type Ig-like domain (D1; aa 28-125) and a membrane proximal C2-type Ig-like domain (D2; aa 135-228) that contains a single glycosylation site (Asn 185). The cytoplasmic domain includes two phosphorylation sites (Tyr 280 and Ser 284) and also has a type II PDZ domain-binding motif (aa 297-299). JAM-A plays a role in epithelial tight junction formation and also regulates monocyte transmigration involved in integrity of epithelial barrier. It participates in the regulation of various biological processes, including paracellular permeability, tight junction formation and maintenance, leukocyte trans-endothelial migration, epithelial-to-mesenchymal transition (EMT), angiogenesis, reovirus binding, and platelet activation. Its dysregulation may contribute to pathological consequences, such as cardiovascular diseases, inflammatory bowel disease, neurological disorders, and various cancer types. In cancer cells, it is not only associated with regulation of cell migration, but it also affects apoptosis, EMT, cancer stem cell maintenance, and cell proliferation. Down-regulation of JAM-A results in elevated cell migration, invasion, and metastasis in various breast cancer cell lines. Its higher levels have been linked to reduced cell migration ability of cancer cells. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Czubak-Prowizor, K., et al. (2022). Mol. Cell. Biochem. 477(1); 79-98; Liu, X., et al. (2020). Nature. 586(7827); 101-107).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service