Skip to Content
Merck
All Photos(15)

Key Documents

ZRB2030

Sigma-Aldrich

Anti-LRRK2/PARK8 Antibody, clone 1C17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Leucine-rich repeat serine/threonine-protein kinase 2;EC:2.7.11.1;Dardarin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C17, recombinant monoclonal

description

1C17 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 286.1 kDa
observed mol wt ~260 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

porcine, monkey, horse, canine, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LRRK2(120892)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C17 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Leucine-rich repeat serine/threonine-protein kinase 2 (LRRK2). It targets an epitope within 25 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the C-terminal region of human Leucine-rich repeat serine/threonine-protein kinase 2 (LRRK2).

Application

Quality Control Testing

Evaluated by Western Blotting in Human kidney tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LRRK2/PARK8 in Human kidney tissue lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LRRK2/PARK8 in Mouse brain tissue lysate.

Affinity Binding Assay: A representative lot of this antibody bound LRRK2/PARK8 peptide with a KD of 8.1 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected LRRK2/PARK8 in A549 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100-1,000 dilution from a representative lot detected LRRK2/PARK8 in human kidney and human Parkinson′s diseased brain tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in Human kidney tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LRRK2/PARK8 in Human kidney tissue lysate.

Target description

Leucine-rich repeat serine/threonine-protein kinase 2 (UniProt: Q5S007; also known as EC:2.7.11.1, Dardarin, LRRK2) is encoded by the LRRK2 (also known as PARK8) gene (Gene ID: 120892) in human. LRRK2 is a homodimeric serine/threonine protein kinase that phosphorylates a broad range of proteins involved in neuronal plasticity, autophagy, and vesicle trafficking. It is expressed in pyramidal neurons in all cortical laminae of the visual cortex, in neurons of the substantia nigra pars compacta, and caudate putamen. It is expressed throughout the adult brain and its expression is particularly high in dopaminoceptive areas of the brain. LRRK2 acts as a key regulator of Rab GTPases. It regulates the GTP/GDP exchange and interaction partners of RABS through phosphorylation. LRRK2 undergoes autophosphorylation and its phosphorylation on serine 910 and either serine 935 or 1444 facilitates its interaction with tyrosine 3-monooxygenase (YWHAG). LRRK2 is also reported to activate a calcium-dependent protein kinase kinase-β (CaMKK-β)/AMPK pathway, which is followed by a persistent increase in autophagosome formation. Overexpression of LRKR2 increases the levels of the autophagy receptor p62 and reduces the number of acidic lysosomes. Mutations in the LRRK2 gene are known to cause autosomal dominant late-onset Parkinson s disease. Overexpression of LRRK2 is reported to increase phosphorylation of amyloid precursor protein (APP) at a residue within the APP intracellular domain (AICD), which promotes AICD transcriptional activity and neurotoxicity in Parkinson s disease leading to a reduction in dopaminergic neurons. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Tolosa, E., et al. (2020). Nat. Rev. Neurol. 16; 97-107; Chen, ZC., et al. (2017). Sci. Signal. 10(488); eaam6790; Guerreiro, PS., et al. (2016). Mol. Neurobiol. 53(5); 3124-3135; Gomez-Suaga, P., et al. (2012). Hum. Mol. Genet. 21(3); 511-525).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service