Skip to Content
Merck
All Photos(1)

Documents

Z719951

Sigma-Aldrich

Gel Repel liquid solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.51

General description

Gel Repel Liquid Solution is a glass plate coating for use with sequencing or PAGE gles and is a non-toxic alternative to silane pre-treatment. Each case contains 4 × 1 oz. vials and instructions for use.

Legal Information

Gel Repel is a trademark of C.B.S. Scientific Co.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F

Flash Point(C)

14.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service