Z664642
Brady® LABXPERT™ tapes
polyester (continuous), W × L 25.40 mm × 9.1 m
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
polyester (continuous)
white
packaging
roll of 9.1 m
manufacturer/tradename
Brady XC-1000-461
W × L
25.40 mm × 9.1 m
Looking for similar products? Visit Product Comparison Guide
Legal Information
Brady is a registered trademark of Brady Worldwide, Inc.
LABXPERT is a trademark of Brady GmbH
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service