Z325163
BRAND® BLAUBRAND® ETERNA volumetric pipette, calibrated To Deliver
capacity 25 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
AR-Glas®
Agency
according to DIN 12 691
description
amber mark: yes
feature
class AS with batch number
packaging
pack of 6 ea
manufacturer/tradename
BRAND 30515
volume measuring range
25 mL, accuracy: 0.03 mL
L
510 mm
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Pipettes come with a detailed batch certificate in each package.
Other Notes
BR30515 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service