Skip to Content
Merck
All Photos(1)

Key Documents

V9752

Sigma-Aldrich

Scorpion venom from Centruroides sculpturatus

Synonym(s):

(Bark Scorpion)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

storage temp.

−20°C

General description

Centruroides sculpturatus scorpion venom induces severe neuromotor disturbance. The venom is composed of prime toxins that target ion-channels especially, the sodium (Na+)-channel.
Scorpion venom are species-specific, which functions in prey capture or predator deterence. It might be lethal to humans. It induces prolonged and excessive firing of neuronal axons, that results in a wide range of signs and symptoms. Scorpion venom might be associated with local pain, bronchoconstriction, neurologic and cardiovascular collapse.

Application

Scorpion venom from Centruroides sculpturatus has been used to activate neuronal sodium channels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen A Klotz et al.
The American journal of medicine, 134(8), 1034-1038 (2021-02-26)
Arizona's rugged desert landscape harbors many venomous animals, including a small nocturnal scorpion, Centruroides sculpturatus, whose venom can cause severe neuromotor disturbance. An effective antivenom is available at selected health care facilities in the state. We analyzed 4398 calls of
Intraspecific variation of Centruroides sculpturatus scorpion venom from two regions of Arizona
Carcamo-Noriega EN, et al.
Archives of Biochemistry and Biophysics, 638, 52-57 (2018)
Neural control of the endocrine rat heart
Jiao JH and Baertschi AJ
Proceedings of the National Academy of Sciences of the USA, 90(16), 7799-7803 (1993)
Norma A Valdez-Cruz et al.
Biochimie, 86(6), 387-396 (2004-09-11)
Current literature concerning the taxonomic names of two possibly distinct species of scorpions from the genus Centruroides (sculpturatus and/or exilicauda) is controversial. This communication reports the results of biochemical, genetic and electrophysiological experiments conducted with C. exilicauda Wood of Baja
Envenomation by the scorpion Centruroides sculpturatus
Curry SC, et al.
Journal of Toxicology: Clinical Toxicology, 21(4-5), 417-449 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service